Learn More
Cayman Chemical L-Kynurenine-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2672568-86-2, 5 mg
SDP
Catalog No. 502912964
Encompass_Preferred
An internal standard for the quantification of kynurenine by GC- or LC-MS
Catalog No. 50-291-2964 Supplier Cayman Chemical Supplier No. 363075 MG
An internal standard for the quantification of kynurenine by GC- or LC-MS