Learn More
Medchemexpress LLC L-kynurenine | 2922-83-0 | MFCD00069912 | 100.0% | 208.21 | C10H12N2O3 | 10 MG
SDP
Catalog No. 5000243352
Encompass_Preferred
L-kynurenine is an analytical reference standard of the endogenous metabolite L-kynurenine, intended for research and analytical applications such as LC/MS calibration, method development, and biochemical assays. It is characterized by CAS 2922-83-0, molecular formula C10H12N2O3, and a molecular weight of 208.21.
  • High purity suitable for analytical and reference use.
  • Known molecular weight for accurate calibration.
  • Solid standard with recommended storage at 4°C, protected from light and moisture.
  • Compatible with LC/MS and other chromatographic or spectrometric methods.
  • Intended for use as an endogenous metabolite and receptor agonist reference.
Catalog No. 50-002-43352 Supplier Medchemexpress LLC Supplier No. HY104026R10MG
L-kynurenine is an analytical reference standard of the endogenous metabolite L-kynurenine, intended for research and analytical applications such as LC/MS calibration, method development, and biochemical assays. It is characterized by CAS 2922-83-0, molecular formula C10H12N2O3, and a molecular weight of 208.21.
  • High purity suitable for analytical and reference use.
  • Known molecular weight for accurate calibration.
  • Solid standard with recommended storage at 4°C, protected from light and moisture.
  • Compatible with LC/MS and other chromatographic or spectrometric methods.
  • Intended for use as an endogenous metabolite and receptor agonist reference.