Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-kynurenine | 2922-83-0 | MFCD00069912 | 100.0% | 208.21 g/mol | C10H12N2O3 | 100 MG
SDP

Catalog No. 5000243456
Encompass_Preferred

L-kynurenine is an analytical reference standard of the tryptophan metabolite L-kynurenine supplied for research and analytical applications. It is used as a calibration and reference material in chromatographic and mass spectrometric methods and is employed in biological studies for its activity as an aryl hydrocarbon receptor agonist.

  • High purity (99.97%) suitable for analytical use.
  • Molecular formula C10H12N2O3 and molecular weight 208.21 g/mol.
  • Intended for HPLC, GC, and MS method development and calibration.
  • Available in small sizes for analytical workflows, including 100 mg.
  • Certificate of analysis and safety data sheet available for quality documentation.

Catalog No. 50-002-43456 Supplier Medchemexpress LLC Supplier No. HY104026R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-kynurenine is an analytical reference standard of the tryptophan metabolite L-kynurenine supplied for research and analytical applications. It is used as a calibration and reference material in chromatographic and mass spectrometric methods and is employed in biological studies for its activity as an aryl hydrocarbon receptor agonist.

  • High purity (99.97%) suitable for analytical use.
  • Molecular formula C10H12N2O3 and molecular weight 208.21 g/mol.
  • Intended for HPLC, GC, and MS method development and calibration.
  • Available in small sizes for analytical workflows, including 100 mg.
  • Certificate of analysis and safety data sheet available for quality documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.