Learn More
Medchemexpress LLC L-kynurenine | 2922-83-0 | MFCD00069912 | 100.0% | 208.21 g/mol | C10H12N2O3 | 5 MG
SDP
Catalog No. 5000243349
Encompass_Preferred
L-kynurenine analytical standard provided as a sealed solid reference material for research and analytical use. It is intended for qualitative and quantitative method development and validation in HPLC, GC, and MS; CAS 2922-83-0; molecular weight 208.21 g/mol.
  • Analytical standard suitable for HPLC, GC, and MS.
  • High purity (99.97%) for accurate quantitation.
  • Solid form, white to light yellow in color.
  • Intended for qualitative and quantitative method development and validation.
  • Store sealed at 4°C, protected from light and moisture.
  • For research use only; not for human consumption or clinical use.
Catalog No. 50-002-43349 Supplier Medchemexpress LLC Supplier No. HY104026R5MG
L-kynurenine analytical standard provided as a sealed solid reference material for research and analytical use. It is intended for qualitative and quantitative method development and validation in HPLC, GC, and MS; CAS 2922-83-0; molecular weight 208.21 g/mol.
  • Analytical standard suitable for HPLC, GC, and MS.
  • High purity (99.97%) for accurate quantitation.
  • Solid form, white to light yellow in color.
  • Intended for qualitative and quantitative method development and validation.
  • Store sealed at 4°C, protected from light and moisture.
  • For research use only; not for human consumption or clinical use.