Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-kynurenine | 2922-83-0 | MFCD00069912 | 100.0% | 208.21 g/mol | C10H12N2O3 | 50 MG
SDP

Catalog No. 5000243502
Encompass_Preferred

L-kynurenine is an analytical reference standard of the endogenous metabolite L-kynurenine, provided as a solid for research and analytical applications. It is used as a reference in qualitative and quantitative assays and method development for HPLC, GC, and MS, and functions biologically as an aryl hydrocarbon receptor agonist. Molecular formula C10H12N2O3; molecular weight 208.21 g/mol.

  • High purity (99.97%).
  • Supplied as a solid suitable for analytical assays.
  • Intended for qualitative and quantitative reference in HPLC, GC, and MS.
  • Molecular weight 208.21 g/mol and formula C10H12N2O3.
  • Store under conditions specified in the certificate of analysis.

Catalog No. 50-002-43502 Supplier Medchemexpress LLC Supplier No. HY104026R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-kynurenine is an analytical reference standard of the endogenous metabolite L-kynurenine, provided as a solid for research and analytical applications. It is used as a reference in qualitative and quantitative assays and method development for HPLC, GC, and MS, and functions biologically as an aryl hydrocarbon receptor agonist. Molecular formula C10H12N2O3; molecular weight 208.21 g/mol.

  • High purity (99.97%).
  • Supplied as a solid suitable for analytical assays.
  • Intended for qualitative and quantitative reference in HPLC, GC, and MS.
  • Molecular weight 208.21 g/mol and formula C10H12N2O3.
  • Store under conditions specified in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.