Learn More
Medchemexpress LLC L-kynurenine | 2922-83-0 | MFCD00069912 | ≥98.0% | 208.21 | C10H12N2O3 | 250 MG
SDP
Catalog No. 5000243455
Encompass_Preferred
L-kynurenine is an endogenous metabolite of L-tryptophan that functions as an agonist of the aryl hydrocarbon receptor (AHR). It is provided for research use as a solid analytical standard for in vitro signaling studies, metabolic profiling, and method development.
  • Acts as an aryl hydrocarbon receptor (AHR) agonist.
  • Suitable as an analytical standard for in vitro and biochemical assays.
  • Available in multiple solid pack sizes, including 250 MG.
  • High purity appropriate for HPLC-based applications (≥98.0%).
  • Molecular formula C10H12N2O3 and molecular weight 208.21.
Catalog No. 50-002-43455 Supplier Medchemexpress LLC Supplier No. HY104026250MG
L-kynurenine is an endogenous metabolite of L-tryptophan that functions as an agonist of the aryl hydrocarbon receptor (AHR). It is provided for research use as a solid analytical standard for in vitro signaling studies, metabolic profiling, and method development.
  • Acts as an aryl hydrocarbon receptor (AHR) agonist.
  • Suitable as an analytical standard for in vitro and biochemical assays.
  • Available in multiple solid pack sizes, including 250 MG.
  • High purity appropriate for HPLC-based applications (≥98.0%).
  • Molecular formula C10H12N2O3 and molecular weight 208.21.