Learn More
Enzo Life Sciences L-Kynurenine (50mg). CAS: 2922-83-0
SDP
Catalog No. 502009553
Encompass_Preferred
Key intermediate in the breakdown pathway of tryptophan. Alternative name: beta-Anthraniloyl-L-alanine, L-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid. Purity: ≥95% (HPLC). Solubility: Soluble in 0.5 M hydrochloric acid (50mg/ml); very poorly soluble in water (0.4g/100ml, 20°C). Long Term Storage: -20°C.
Catalog No. 50-200-9553 Supplier Enzo Life Sciences Supplier No. ALX550408M050
Key intermediate in the breakdown pathway of tryptophan. Alternative name: beta-Anthraniloyl-L-alanine, L-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid. Purity: ≥95% (HPLC). Solubility: Soluble in 0.5 M hydrochloric acid (50mg/ml); very poorly soluble in water (0.4g/100ml, 20°C). Long Term Storage: -20°C.