Learn More
Cayman Chemical L,L-Dityrosine, Purity: ≥90%, CAS: 63442-81-9, 5 mg
SDP
Catalog No. 502910793
Encompass_Preferred
An amino acid conjugate; cross-links of it can form between fibrinogen molecules in the presence of hematin and plasma-generated hydrogen peroxide under non-thermal plasma exposure; cross-links of it can form between human amylin peptides in the presence of copper (II) ions and hydrogen peroxide
Catalog No. 50-291-0793 Supplier Cayman Chemical Supplier No. 357465 MG
An amino acid conjugate; cross-links of it can form between fibrinogen molecules in the presence of hematin and plasma-generated hydrogen peroxide under non-thermal plasma exposure; cross-links of it can form between human amylin peptides in the presence of copper (II) ions and hydrogen peroxide