Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L,L-Dityrosine hydrochloride | 221308-01-6 | 99.9% | 433.28 | C18H22Cl2N2O6 | 1 ML
SDP

Catalog No. 5000248950
Encompass_Preferred

L,L-Dityrosine hydrochloride is an amino acid derivative present in acid hydrolysates of biological materials, including insect cuticular resilin. It is supplied in solution and solid forms for research and analytical use, with documented molecular properties and high purity suitable for biochemical assays.

  • Purity 99.91%.
  • Molecular weight 433.28.
  • Molecular formula C18H22Cl2N2O6.
  • Available as a 10 mM solution in DMSO and as multiple solid sizes.
  • Intended for biochemical assays and analytical reference use.
  • H2O listed as an optional solvent for stock solutions.

Catalog No. 50-002-48950 Supplier Medchemexpress LLC Supplier No. HY101552B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L,L-Dityrosine hydrochloride is an amino acid derivative present in acid hydrolysates of biological materials, including insect cuticular resilin. It is supplied in solution and solid forms for research and analytical use, with documented molecular properties and high purity suitable for biochemical assays.

  • Purity 99.91%.
  • Molecular weight 433.28.
  • Molecular formula C18H22Cl2N2O6.
  • Available as a 10 mM solution in DMSO and as multiple solid sizes.
  • Intended for biochemical assays and analytical reference use.
  • H2O listed as an optional solvent for stock solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.