Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L,L-dityrosine (hydrochloride) | 221308-01-6 | 99.9% | 433.28 | C18H22Cl2N2O6 | 5 MG
SDP

Catalog No. 5000247996
Encompass_Preferred

L,L-dityrosine hydrochloride (CAS 221308-01-6) is a dipeptide-derived biochemical constituent of acid hydrolysates of biological materials, including insect cuticular resilin. It is supplied for research and analytical applications and is available as a high-purity solid or as a DMSO solution for experimental flexibility.

  • High purity (>99.9%) suitable for analytical work
  • Well-characterized molecular properties: MW 433.28, formula C18H22Cl2N2O6
  • Available as small solid quantities and as a 10 mM DMSO solution
  • Useful for studies of protein crosslinking and oxidative modifications
  • Provided with a safety data sheet for handling and storage guidance

Catalog No. 50-002-47996 Supplier Medchemexpress LLC Supplier No. HY101552B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L,L-dityrosine hydrochloride (CAS 221308-01-6) is a dipeptide-derived biochemical constituent of acid hydrolysates of biological materials, including insect cuticular resilin. It is supplied for research and analytical applications and is available as a high-purity solid or as a DMSO solution for experimental flexibility.

  • High purity (>99.9%) suitable for analytical work
  • Well-characterized molecular properties: MW 433.28, formula C18H22Cl2N2O6
  • Available as small solid quantities and as a 10 mM DMSO solution
  • Useful for studies of protein crosslinking and oxidative modifications
  • Provided with a safety data sheet for handling and storage guidance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.