Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L,L-dityrosine hydrochloride | 221308-01-6 | MFCD22575199 | 99.9% | 433.28 g/mol | C18H22Cl2N2O6 | 10 MG
SDP

Catalog No. 5000248772
Encompass_Preferred

L,L-dityrosine hydrochloride is the hydrochloride salt of dityrosine used as a research reagent and analytical reference standard in biochemical and materials studies. It is typically supplied as a light yellow to light brown solid and is used for assay calibration and structural studies.

  • High purity (99.9%)
  • Light yellow to light brown solid appearance
  • Water solubility: 3.33 mg/mL with ultrasonic warming to 60°C
  • Molecular weight: 433.28 g/mol
  • Store solid at 4°C, sealed and protected from moisture
  • Available in small pack sizes suitable for research use

Catalog No. 50-002-48772 Supplier Medchemexpress LLC Supplier No. HY101552B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L,L-dityrosine hydrochloride is the hydrochloride salt of dityrosine used as a research reagent and analytical reference standard in biochemical and materials studies. It is typically supplied as a light yellow to light brown solid and is used for assay calibration and structural studies.

  • High purity (99.9%)
  • Light yellow to light brown solid appearance
  • Water solubility: 3.33 mg/mL with ultrasonic warming to 60°C
  • Molecular weight: 433.28 g/mol
  • Store solid at 4°C, sealed and protected from moisture
  • Available in small pack sizes suitable for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.