Learn More
Medchemexpress LLC L,L-dityrosine hydrochloride | 221308-01-6 | MFCD22575199 | 99.9% | 433.28 g/mol | C18H22Cl2N2O6 | 50 MG
SDP
Catalog No. 5000248268
Encompass_Preferred
L,L-Dityrosine hydrochloride is a small-molecule reference compound occurring in acid hydrolysates of biological materials and used in biochemical research. Supplied as a solid, it is typically employed as an analytical standard in studies of protein cross-linking, oxidative modification, and mass spectrometry workflows.
  • High purity (≈99.9%) suitable for analytical use.
  • Molecular weight 433.28 g/mol.
  • Solid, light yellow to light brown appearance.
  • Available in multiple pack sizes, including 50 mg.
  • Commonly used as a reference standard for biochemical assays and mass spectrometry.
Catalog No. 50-002-48268 Supplier Medchemexpress LLC Supplier No. HY101552B50MG
L,L-Dityrosine hydrochloride is a small-molecule reference compound occurring in acid hydrolysates of biological materials and used in biochemical research. Supplied as a solid, it is typically employed as an analytical standard in studies of protein cross-linking, oxidative modification, and mass spectrometry workflows.
  • High purity (≈99.9%) suitable for analytical use.
  • Molecular weight 433.28 g/mol.
  • Solid, light yellow to light brown appearance.
  • Available in multiple pack sizes, including 50 mg.
  • Commonly used as a reference standard for biochemical assays and mass spectrometry.