Learn More
Medchemexpress LLC L,L-dityrosine (o,o'-dityrosine) | 63442-81-9 | 99.8% | 360.36 g/mol | C18H20N2O6 | 10 MG
SDP
Catalog No. 5000245469
Encompass_Preferred
L,L-Dityrosine (o,o'-dityrosine) is a dityrosine-linked amino acid derivative used as a biochemical reference standard and reagent in studies of protein crosslinking, oxidative modification, and structural biology. Supplied as a high-purity solid for laboratory research use only.
  • High purity suitable for analytical and research applications.
  • Used in studies of protein crosslinking and oxidative damage.
  • Provided as a stable solid convenient for storage and handling.
  • Characterized by CAS 63442-81-9 and known molecular weight.
  • Supporting documentation available: datasheet, COA, and SDS.
Catalog No. 50-002-45469 Supplier Medchemexpress LLC Supplier No. HY101552A10MG
L,L-Dityrosine (o,o'-dityrosine) is a dityrosine-linked amino acid derivative used as a biochemical reference standard and reagent in studies of protein crosslinking, oxidative modification, and structural biology. Supplied as a high-purity solid for laboratory research use only.
  • High purity suitable for analytical and research applications.
  • Used in studies of protein crosslinking and oxidative damage.
  • Provided as a stable solid convenient for storage and handling.
  • Characterized by CAS 63442-81-9 and known molecular weight.
  • Supporting documentation available: datasheet, COA, and SDS.