Learn More
Medchemexpress LLC L,L-dityrosine (o,o'-dityrosine) | 63442-81-9 | 99.8% | 360.37 | C18H20N2O6 | 50 MG
SDP
Catalog No. 5000246556
Encompass_Preferred
L,L-Dityrosine (o,o'-Dityrosine) is a dimeric amino-acid derivative used as a research reagent and analytical standard for studies of protein crosslinking, oxidative modifications, and metabolite analysis. Supplied as a high-purity solid, it is intended for laboratory research and analytical method development; proper storage ensures stability and reproducible results.
  • High purity (99.8%) suitable for analytical work.
  • Useful as an analytical standard and reference material.
  • Applicable to studies of protein crosslinking and oxidative modifications.
  • Available in small pack sizes for laboratory experiments.
  • Store sealed at 4°C; in solution, keep at -80°C (up to 6 months) or -20°C (short term).
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-46556 Supplier Medchemexpress LLC Supplier No. HY101552A50MG
L,L-Dityrosine (o,o'-Dityrosine) is a dimeric amino-acid derivative used as a research reagent and analytical standard for studies of protein crosslinking, oxidative modifications, and metabolite analysis. Supplied as a high-purity solid, it is intended for laboratory research and analytical method development; proper storage ensures stability and reproducible results.
  • High purity (99.8%) suitable for analytical work.
  • Useful as an analytical standard and reference material.
  • Applicable to studies of protein crosslinking and oxidative modifications.
  • Available in small pack sizes for laboratory experiments.
  • Store sealed at 4°C; in solution, keep at -80°C (up to 6 months) or -20°C (short term).
  • For research use only; not for human therapeutic use.