Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L,L-dityrosine (o,o'-dityrosine) | 63442-81-9 | MFCD32201023 | 99.8% | 360.36 g/mol | C18H20N2O6 | 5 MG
SDP

Catalog No. 5000248474
Encompass_Preferred

L,L-dityrosine (o,o'-dityrosine) is a dimer of the amino acid tyrosine found in acid hydrolysates of biological materials and reported elevated in certain neurochemical models. Supplied as a research-grade reference standard, it is intended for biochemical, analytical, and chromatographic studies where a well-characterized dityrosine reference is required.

  • High purity (99.80%) for reliable analytical results.
  • Molecular formula C18H20N2O6; molecular weight 360.36 g/mol.
  • Supplied as a dry, white to off-white solid suitable for dissolution in common solvents.
  • Used as a reference standard and biomarker in oxidative stress and protein crosslinking studies.
  • Accompanied by a datasheet and certificate of analysis for batch-specific quality data.

Catalog No. 50-002-48474 Supplier Medchemexpress LLC Supplier No. HY101552A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L,L-dityrosine (o,o'-dityrosine) is a dimer of the amino acid tyrosine found in acid hydrolysates of biological materials and reported elevated in certain neurochemical models. Supplied as a research-grade reference standard, it is intended for biochemical, analytical, and chromatographic studies where a well-characterized dityrosine reference is required.

  • High purity (99.80%) for reliable analytical results.
  • Molecular formula C18H20N2O6; molecular weight 360.36 g/mol.
  • Supplied as a dry, white to off-white solid suitable for dissolution in common solvents.
  • Used as a reference standard and biomarker in oxidative stress and protein crosslinking studies.
  • Accompanied by a datasheet and certificate of analysis for batch-specific quality data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.