Learn More
MOLECULAR DEPOT LLC L-lactate - 1 mL
Catalog No. 5000013542
Encompass_Preferred
Lactate for Interference Testing is a highly concentrated solution of L-Lactate (100 mg/mL) provided with the corresponding diluent This solution can be spiked into samples to test Lactate (L-Lactate) interference in assays For in vitro research use only
Catalog No. 50-000-13542 Supplier MOLECULAR DEPOT LLC Supplier No. B2010200
Lactate for Interference Testing is a highly concentrated solution of L-Lactate (100 mg/mL) provided with the corresponding diluent This solution can be spiked into samples to test Lactate (L-Lactate) interference in assays For in vitro research use only