Learn More
Medchemexpress LLC L-Lactate-13C sodium | 201595-69-9 | 98.0% | 113.05 | 5 MG
SDP
Catalog No. 5000393350
Encompass_Preferred
L-Lactate-13C sodium is the 13C-labeled version of L-Lactic acid sodium. It serves as a building block and can be utilized as a precursor for the production of the bioplastic polymer poly-lactic acid. L-Lactic acid sodium also exhibits antiproliferative activity. This compound can be used as a tracer and an internal standard for quantitative analysis. Stable heavy isotopes, including carbon-13, are incorporated into drug molecules as tracers in drug development, with deuteration noted for its potential impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs (in vitro)
Catalog No. 50-003-93350 Supplier Medchemexpress LLC Supplier No. 5MGHYY0479AS2
L-Lactate-13C sodium is the 13C-labeled version of L-Lactic acid sodium. It serves as a building block and can be utilized as a precursor for the production of the bioplastic polymer poly-lactic acid. L-Lactic acid sodium also exhibits antiproliferative activity. This compound can be used as a tracer and an internal standard for quantitative analysis. Stable heavy isotopes, including carbon-13, are incorporated into drug molecules as tracers in drug development, with deuteration noted for its potential impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs (in vitro)