Learn More
Medchemexpress LLC L-lactate-13C (sodium) | 201595-69-9 | 98.0% | 113.05 | C213CH5NaO3 | 50mg
SDP
Catalog No. 5000269711
Encompass_Preferred
L-Lactate-13C ((S)-2-Hydroxypropanoic acid-13C) sodium is the 13C-labeled L-Lactic acid sodium (HY-W040233) L-lactate Sodium is a buildiing block which can be used as a precursor for the production of the bioplastic polymer poly-lactic acid L-Lactic acid Sodium has antiproliferative activity[1][2][3]
Catalog No. 50-002-69711 Supplier Medchemexpress LLC Supplier No. HYY0479AS250MG
L-Lactate-13C ((S)-2-Hydroxypropanoic acid-13C) sodium is the 13C-labeled L-Lactic acid sodium (HY-W040233) L-lactate Sodium is a buildiing block which can be used as a precursor for the production of the bioplastic polymer poly-lactic acid L-Lactic acid Sodium has antiproliferative activity[1][2][3]