Learn More
Medchemexpress LLC L-lactate-13C (sodium) | 201595-71-3 | 99.9% | 115.04 | 13C3H5NaO3 | 50mg
SDP
Catalog No. 5000269213
Encompass_Preferred
L-Lactic acid-13C3 ((S)-2-hydroxypropanoic-13C3) sodium (20% in water) is the 13C labeled L-Lactic acid L-Lactic acid-13C3 sodium (20% in water) can be used for lactate metabolism research[1]
Catalog No. 50-002-69213 Supplier Medchemexpress LLC Supplier No. HYY0479AS50MG
L-Lactic acid-13C3 ((S)-2-hydroxypropanoic-13C3) sodium (20% in water) is the 13C labeled L-Lactic acid L-Lactic acid-13C3 sodium (20% in water) can be used for lactate metabolism research[1]