Learn More
ETON BIOSCIENCE INC L-LACTATE ASSAY KIT 5ML
Catalog No. NC0014474
Encompass
Lactate dehydrogenase converts lactate and NAD+ into pyruvate and NADH. The Lactate Assay Kit is based on the reduction of the terazolium salt INT in a NADH-coupled enzyme reaction to formazan, which exhibits an absorbance maximum at 490 nm.
Catalog No. NC0014474 Supplier ETON BIOSCIENCE INC Supplier No. 1200011002
Lactate dehydrogenase converts lactate and NAD+ into pyruvate and NADH. The Lactate Assay Kit is based on the reduction of the terazolium salt INT in a NADH-coupled enzyme reaction to formazan, which exhibits an absorbance maximum at 490 nm.