Learn More
Sigma Aldrich Fine Chemicals Biosciences L Lactate Assay Kit suffic
Catalog No. 502904603
Encompass
Lactate is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions. Monitoring lactate levels is therefore a good indicator of the balance between tissue oxygen demand and utilization and is useful when studying cellular and animal physiology. Simple direct and automation-ready procedures for measuring lactate concentration are very desirable.
Catalog No. 50-290-4603 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3291KT
Lactate is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions. Monitoring lactate levels is therefore a good indicator of the balance between tissue oxygen demand and utilization and is useful when studying cellular and animal physiology. Simple direct and automation-ready procedures for measuring lactate concentration are very desirable.