Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Lactate Dehydrogenase L-
Catalog No. 502901127
Encompass
L-lactateNAD oxidoreductase.L-lactate dehydrogenase catalyzes the reversible reduction of pyruvate to L-lactate.
Catalog No. 50-290-1127 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10127876001
L-lactateNAD oxidoreductase.L-lactate dehydrogenase catalyzes the reversible reduction of pyruvate to L-lactate.