Learn More
Sigma Aldrich Fine Chemicals Biosciences L Lactate Dehydrogenase L
Catalog No. 502905103
Encompass
L-lactateNAD oxidoreductase.L-lactate dehydrogenase catalyzes the reversible reduction of pyruvate to L-lactate.
Catalog No. 50-290-5103 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10127884001
L-lactateNAD oxidoreductase.L-lactate dehydrogenase catalyzes the reversible reduction of pyruvate to L-lactate.