Learn More
YSI 3P LifeScience 1530 L-Lactate Standard, 30 mmol/L (125 mL)
Catalog No. 50200109
Encompass
30 mmol/L L-Lactate standard solution for use with YSI 1500, 2300, 2700, 2900 Series and 7100 Analyzers.
Catalog No. 50-200-109 Supplier YSI 3P LifeScience Supplier No. 1530
30 mmol/L L-Lactate standard solution for use with YSI 1500, 2300, 2700, 2900 Series and 7100 Analyzers.