Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Lactic acid lithium | 27848-80-2 | 98.0% | 10 G
SDP

Catalog No. 5000410620
Encompass_Preferred

L-Lactic acid lithium is a lithium salt of L-Lactic acid with bioconductive properties. It serves as a precursor for the production of bioplastic polymer polylactic acid.

  • Lithium salt of L-lactic acid
  • Precursor for bioplastic polymer polylactic acid production
  • Molecular weight: 96.01
  • Chemical formula: C3H5LiO3
  • Appearance: solid
  • Color: white to off-white
  • Storage: room temperature, dry and cool

Catalog No. 50-004-10620 Supplier Medchemexpress LLC Supplier No. HYD088410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Lactic acid lithium is a lithium salt of L-Lactic acid with bioconductive properties. It serves as a precursor for the production of bioplastic polymer polylactic acid.

  • Lithium salt of L-lactic acid
  • Precursor for bioplastic polymer polylactic acid production
  • Molecular weight: 96.01
  • Chemical formula: C3H5LiO3
  • Appearance: solid
  • Color: white to off-white
  • Storage: room temperature, dry and cool

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.