Learn More
Medchemexpress LLC L-lactic acid (S)-2-hydroxypropanoic acid | 79-33-4 | MFCD00004520 | 99.6% | 90.08 g·mol⁻1 | C3H6O3 | 5g
SDP
Catalog No. 5000268240
Encompass_Preferred
L-Lactic acid is a buildiing block which can be used as a precursor for the production of the bioplastic polymer poly-lactic acid L-Lactic acid has antiproliferative activity[1][2][3]
Catalog No. 50-002-68240 Supplier Medchemexpress LLC Supplier No. HYY04795G
L-Lactic acid is a buildiing block which can be used as a precursor for the production of the bioplastic polymer poly-lactic acid L-Lactic acid has antiproliferative activity[1][2][3]