Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich L-(+)-Lactic Acid Solution

Catalog No. 27714500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-714-500ML 500 mL
27-714-1L 1 L
27-714-25L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 27-714-500ML Supplier Sigma Aldrich Supplier No. 27714500ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The article number 27714-4X2.5L will be discontinued. Please order the single bottle 27714-2.5L which is physically identical with the same exact specifications.The article number 27714-6X1L will be discontinued. Please order the single bottle 27714-1L which is physically identical with the same exact specifications.

Specifications

CAS 79-33-4
Molecular Formula C3H6O3
Linear Formula C3H6O3
MDL Number MFCD00064266
Quantity 500 mL
Synonym (S)-2-Hydroxypropionic acid; Sarcolactic acid
Molecular Weight (g/mol) 90.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.