Learn More
Cayman Chemical L-Leu-AMC (hydrochloride), Purity: ≥98%, CAS: 62480-44-8, 100 mg
SDP
Catalog No. 502950180
Encompass_Preferred
A fluorogenic substrate for leucine aminopeptidase; AMC is released upon enzymatic cleavage by leucine aminopeptidase and its fluorescence can be used to quantify leucine aminopeptidase activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-295-0180 Supplier Cayman Chemical Supplier No. 34267100 MG
A fluorogenic substrate for leucine aminopeptidase; AMC is released upon enzymatic cleavage by leucine aminopeptidase and its fluorescence can be used to quantify leucine aminopeptidase activity; ex/em = 340-360/440-460 nm, respectively