Learn More
Cayman Chemical L-Leucine-d10, Purity: ≥99% deuterated forms (d1-d10), CAS: 106972-44-5, 25 mg
SDP
Catalog No. 502911042
Encompass_Preferred
An internal standard for the quantification of L-leucine by GC- or LC-MS
Catalog No. 50-291-1042 Supplier Cayman Chemical Supplier No. 3484325 MG
An internal standard for the quantification of L-leucine by GC- or LC-MS