Learn More
Medchemexpress LLC L-Leucinamide, N-acetyl-L-leucyl-N-[(1S)-1-formylpentyl]- | 110044-82-1 | ≥98.0% | 383.53 | 100 MG
SDP
Catalog No. 5000385191
Encompass_Preferred
MG-101 (ALLN) is an inhibitor of cysteine proteases, specifically inhibiting calpain I, calpain II, cathepsin B, and cathepsin L with Kis of 190, 220, 150, and 500 pM, respectively. It induces apoptosis and inhibits tumor growth, making it suitable for research on colon cancer. For research use only.
  • Inhibits cysteine proteases: calpain I, calpain II, cathepsin B, and cathepsin L
  • Induces apoptosis
  • Inhibits tumor growth
  • Can be used for colon cancer research
Catalog No. 50-003-85191 Supplier Medchemexpress LLC Supplier No. HY18964100MG
MG-101 (ALLN) is an inhibitor of cysteine proteases, specifically inhibiting calpain I, calpain II, cathepsin B, and cathepsin L with Kis of 190, 220, 150, and 500 pM, respectively. It induces apoptosis and inhibits tumor growth, making it suitable for research on colon cancer. For research use only.
  • Inhibits cysteine proteases: calpain I, calpain II, cathepsin B, and cathepsin L
  • Induces apoptosis
  • Inhibits tumor growth
  • Can be used for colon cancer research