Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Leucine-1-13C,15N | 80134-83-4 | 133.16 | 100 MG
SDP

Catalog No. 5000389794
Encompass_Preferred

L-Leucine-1-13C,15N is a form of L-Leucine that has been labeled with 13C and 15N isotopes. L-Leucine is an essential branched-chain amino acid (BCAA) known to activate the mTOR signaling pathway. This compound is intended for research use only.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 99.93%.
  • Appearance: Solid.
  • Color: White to off-white.

Catalog No. 50-003-89794 Supplier Medchemexpress LLC Supplier No. HYN0486S7100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Leucine-1-13C,15N is a form of L-Leucine that has been labeled with 13C and 15N isotopes. L-Leucine is an essential branched-chain amino acid (BCAA) known to activate the mTOR signaling pathway. This compound is intended for research use only.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 99.93%.
  • Appearance: Solid.
  • Color: White to off-white.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.