Learn More
Medchemexpress LLC L-Leucine-1-13C,15N | 80134-83-4 | 133.16 | 25 MG
SDP
Catalog No. 5000390341
Encompass_Preferred
L-Leucine-1-13C,15N is a 13C- and 15N-labeled form of L-Leucine, an essential branched-chain amino acid (BCAA). This compound is known to activate the mTOR signaling pathway and is intended solely for research applications.
  • For research use only
  • Activates the mTOR signaling pathway
  • 13C- and 15N-labeled L-Leucine
  • Essential branched-chain amino acid
Catalog No. 50-003-90341 Supplier Medchemexpress LLC Supplier No. 25MGHYN0486S7
Add to Cart
Add to Cart
L-Leucine-1-13C,15N is a 13C- and 15N-labeled form of L-Leucine, an essential branched-chain amino acid (BCAA). This compound is known to activate the mTOR signaling pathway and is intended solely for research applications.
  • For research use only
  • Activates the mTOR signaling pathway
  • 13C- and 15N-labeled L-Leucine
  • Essential branched-chain amino acid