Learn More
Medchemexpress LLC L-Leucine-1-13C,15N | 80134-83-4 | 133.16 | 50 MG
SDP
Catalog No. 5000389242
Encompass_Preferred
L-Leucine-1-13C,15N is the 13C- and 15N-labeled L-Leucine. L-Leucine is an essential branched-chain amino acid (BCAA), which activates the mTOR signaling pathway. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Activates the mTOR signaling pathway.
  • 13C- and 15N-labeled L-Leucine.
  • Essential branched-chain amino acid (BCAA).
Catalog No. 50-003-89242 Supplier Medchemexpress LLC Supplier No. HYN0486S750MG
L-Leucine-1-13C,15N is the 13C- and 15N-labeled L-Leucine. L-Leucine is an essential branched-chain amino acid (BCAA), which activates the mTOR signaling pathway. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Activates the mTOR signaling pathway.
  • 13C- and 15N-labeled L-Leucine.
  • Essential branched-chain amino acid (BCAA).