Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-leucine-1-13C,15N | 80134-83-4 | 98.0% | 133.16 g/mol | C5(13C)H13(15N)O2 | 10 MG
SDP

Catalog No. 5000295895
Encompass_Preferred

L-leucine labeled with 13C at the 1-position and 15N, supplied as a stable-isotope tracer for research use. It is intended for use as a tracer or internal standard in analytical methods such as NMR, GC-MS, and LC-MS, and should be stored sealed away from moisture and light.

  • Isotopically labeled for tracer and internal standard applications.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Purity 98.0% as listed.
  • Available in small mg quantities for research workflows.
  • Store at 4°C sealed; in solvent, store -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-95895 Supplier Medchemexpress LLC Supplier No. HYN0486S710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-leucine labeled with 13C at the 1-position and 15N, supplied as a stable-isotope tracer for research use. It is intended for use as a tracer or internal standard in analytical methods such as NMR, GC-MS, and LC-MS, and should be stored sealed away from moisture and light.

  • Isotopically labeled for tracer and internal standard applications.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Purity 98.0% as listed.
  • Available in small mg quantities for research workflows.
  • Store at 4°C sealed; in solvent, store -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.