Learn More
Medchemexpress LLC L-leucine-13C6, 15N | 202406-52-8 | 99.9% | 138.12 g/mol | 13C6H1315NO2 | 10 MG
SDP
Catalog No. 5000295873
Encompass_Preferred
L-leucine-13C6,15N is the 13C- and 15N-labeled form of the essential branched-chain amino acid L-leucine, supplied as a high-purity stable isotope powder for use as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS studies. The material supports metabolic and pharmacokinetic research, including studies of mTOR signaling, and has defined storage recommendations for powder and solutions.
  • 13C6 and 15N labeled leucine.
  • High purity (99.9%).
  • Supplied as a powder suitable for tracer and quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Available in small research sizes (e.g., 1 mg, 5 mg, 10 mg).
  • Storage: powder at -20°C for long-term; in solvent at -80°C for extended storage.
Catalog No. 50-002-95873 Supplier Medchemexpress LLC Supplier No. HYN0486S810MG
L-leucine-13C6,15N is the 13C- and 15N-labeled form of the essential branched-chain amino acid L-leucine, supplied as a high-purity stable isotope powder for use as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS studies. The material supports metabolic and pharmacokinetic research, including studies of mTOR signaling, and has defined storage recommendations for powder and solutions.
  • 13C6 and 15N labeled leucine.
  • High purity (99.9%).
  • Supplied as a powder suitable for tracer and quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Available in small research sizes (e.g., 1 mg, 5 mg, 10 mg).
  • Storage: powder at -20°C for long-term; in solvent at -80°C for extended storage.