Learn More
Cambridge Isotope Laboratories L-Leucine (13C6, 99%), 61-90-5, MFCD00002617, 0.25G
Catalog No. NC0991332
Encompass
Chemical Formula: (*CH3)2*CH*CH2*CH(NH2)*COOH, Molecular Weight: 137.13, Synonym: (S)-2-Amino-4-methylpentanoic acid-13C6, 13C Labeled L-leucine. For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.
Catalog No. NC0991332 Supplier Cambridge Isotope Laboratories Supplier No. CLM2262H0.25
Chemical Formula: (*CH3)2*CH*CH2*CH(NH2)*COOH, Molecular Weight: 137.13, Synonym: (S)-2-Amino-4-methylpentanoic acid-13C6, 13C Labeled L-leucine. For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.