Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Leucine-18O2 | 73579-45-0 | 135.17 | 25 MG
SDP

Catalog No. 5000390241
Encompass_Preferred

L-Leucine-18O2 is an 18O-labeled L-Leucine, an essential branched-chain amino acid (BCAA) known for activating the mTOR signaling pathway. It is suitable for various research applications requiring isotopic labeling.

  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Purity: 98%
  • Appearance: Solid, white to off-white
  • For research use only

Catalog No. 50-003-90241 Supplier Medchemexpress LLC Supplier No. 25MGHYN0486S10
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Leucine-18O2 is an 18O-labeled L-Leucine, an essential branched-chain amino acid (BCAA) known for activating the mTOR signaling pathway. It is suitable for various research applications requiring isotopic labeling.

  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Purity: 98%
  • Appearance: Solid, white to off-white
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.