Learn More
Medchemexpress LLC L-Leucine-2-13C 15N 100mg
SDP
Catalog No. 5000669457
Encompass_Preferred
L-Leucine-2-13C 15N is the 13C- and 15N-labeled L-Leucine L-Leucine is an essential branched-chain amino acid (BCAA) which activates the mTOR signaling pathway[1]
Catalog No. 50-006-69457 Supplier Medchemexpress LLC Supplier No. 100MGHYN0486S6
L-Leucine-2-13C 15N is the 13C- and 15N-labeled L-Leucine L-Leucine is an essential branched-chain amino acid (BCAA) which activates the mTOR signaling pathway[1]