Learn More
Medchemexpress LLC L-leucine-2,3,3,4,5,5,5,5',5',5'-d10, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 1190594-22-9 | 98.9% | 363.47 g/mol | C21H13D10NO4 | 5 MG
SDP
Catalog No. 5000213248
Encompass_Preferred
Fmoc-leucine-d10 is a deuterium-labeled N-(9-fluorenylmethoxycarbonyl) derivative of L-leucine supplied as a stable isotope reagent for analytical and research use. It is typically used as an internal standard for mass spectrometry, for isotope tracing, and in labeled-synthesis applications.
  • Deuterium-labeled (d10) Fmoc-protected leucine.
  • Suitable as an internal standard for mass spectrometry.
  • Useful for isotope tracing and labeled-synthesis studies.
  • High purity: 98.9%.
  • Molecular weight: 363.47 g/mol.
  • Supplied in small research quantities (5 mg).
Catalog No. 50-002-13248 Supplier Medchemexpress LLC Supplier No. HY101064S35MG
Fmoc-leucine-d10 is a deuterium-labeled N-(9-fluorenylmethoxycarbonyl) derivative of L-leucine supplied as a stable isotope reagent for analytical and research use. It is typically used as an internal standard for mass spectrometry, for isotope tracing, and in labeled-synthesis applications.
  • Deuterium-labeled (d10) Fmoc-protected leucine.
  • Suitable as an internal standard for mass spectrometry.
  • Useful for isotope tracing and labeled-synthesis studies.
  • High purity: 98.9%.
  • Molecular weight: 363.47 g/mol.
  • Supplied in small research quantities (5 mg).