Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-leucine-2,3,3,4,5,5,5,5',5',5'-d10, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 1190594-22-9 | None | 98.9% | 363.47 | C21H13D10NO4 | 25 MG
SDP

Catalog No. 5000213247
Encompass_Preferred

Fmoc-leucine-d10 is a deuterium-labeled, Fmoc-protected L-leucine supplied as a research reagent for isotopic labeling, peptide synthesis, and analytical standards. It is offered at high chemical purity for use in mass spectrometry, synthetic chemistry, and biological studies where isotopic tracers are required.

  • Deuterium-labeled (d10) Fmoc-protected amino acid.
  • High purity: 98.9%.
  • Molecular weight 363.47 and formula C21H13D10NO4.
  • Available in small research quantities such as 25 MG.
  • Suitable as an MS internal standard and isotopic tracer.
  • Reported to modulate PPARγ activity in biological assays.

Catalog No. 50-002-13247 Supplier Medchemexpress LLC Supplier No. HY101064S325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-leucine-d10 is a deuterium-labeled, Fmoc-protected L-leucine supplied as a research reagent for isotopic labeling, peptide synthesis, and analytical standards. It is offered at high chemical purity for use in mass spectrometry, synthetic chemistry, and biological studies where isotopic tracers are required.

  • Deuterium-labeled (d10) Fmoc-protected amino acid.
  • High purity: 98.9%.
  • Molecular weight 363.47 and formula C21H13D10NO4.
  • Available in small research quantities such as 25 MG.
  • Suitable as an MS internal standard and isotopic tracer.
  • Reported to modulate PPARγ activity in biological assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.