Learn More
Medchemexpress LLC L-Leucine-5,5,5-d3 | 87828-86-2 | 134.19 | 25 MG
SDP
Catalog No. 5000390244
Encompass_Preferred
L-Leucine-d3 is the deuterium labeled L-Leucine, an essential branched-chain amino acid (BCAA) that activates the mTOR signaling pathway. It can be used as a tracer and as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs. This product is for research use only.
  • Used as a tracer in research
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterated for potential pharmacokinetic and metabolic profile studies
Catalog No. 50-003-90244 Supplier Medchemexpress LLC Supplier No. 25MGHYN0486S9
L-Leucine-d3 is the deuterium labeled L-Leucine, an essential branched-chain amino acid (BCAA) that activates the mTOR signaling pathway. It can be used as a tracer and as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs. This product is for research use only.
  • Used as a tracer in research
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterated for potential pharmacokinetic and metabolic profile studies