Learn More
Medchemexpress LLC L-leucine-5,5,5-d3 | 87828-86-2 | 99.9% | 134.19 g/mol | C6H10D3NO2 | 10 MG
SDP
Catalog No. 5000296372
Encompass_Preferred
L-leucine-d3 is the deuterium-labeled form of the essential branched-chain amino acid L-leucine, provided as a stable isotope reagent for research applications such as metabolic tracing and mass spectrometry. It features high isotopic enrichment and is for research use only.
  • Deuterium labeled at the methyl group (5,5,5-d3).
  • High purity, reported as 99.93% (rounded to 99.9%).
  • CAS number 87828-86-2 for chemical identification.
  • Suitable for use in mass spectrometry and metabolic tracing experiments.
  • Available in small research pack sizes, including 10 mg.
Catalog No. 50-002-96372 Supplier Medchemexpress LLC Supplier No. HYN0486S910MG
L-leucine-d3 is the deuterium-labeled form of the essential branched-chain amino acid L-leucine, provided as a stable isotope reagent for research applications such as metabolic tracing and mass spectrometry. It features high isotopic enrichment and is for research use only.
  • Deuterium labeled at the methyl group (5,5,5-d3).
  • High purity, reported as 99.93% (rounded to 99.9%).
  • CAS number 87828-86-2 for chemical identification.
  • Suitable for use in mass spectrometry and metabolic tracing experiments.
  • Available in small research pack sizes, including 10 mg.