Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Leucine-5,5,5-d3, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 538372-74-6 | 99.25% | 356.43 | 25 MG
SDP

Catalog No. 5000346777
Encompass_Preferred

Fmoc-leucine-d3 is a white to off-white solid that has been tested and complies with given specifications. It is consistent with structure based on 1H NMR Spectrum and LCMS analysis.

  • Appearance: white to off-white solid
  • 1H NMR Spectrum: consistent with structure
  • LCMS: consistent with structure
  • Isotopic enrichment: 99.86%
  • Storage: powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-46777 Supplier Medchemexpress LLC Supplier No. HY101064S250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-leucine-d3 is a white to off-white solid that has been tested and complies with given specifications. It is consistent with structure based on 1H NMR Spectrum and LCMS analysis.

  • Appearance: white to off-white solid
  • 1H NMR Spectrum: consistent with structure
  • LCMS: consistent with structure
  • Isotopic enrichment: 99.86%
  • Storage: powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.