Learn More
AdipoGen L-Leucine-7-amido-4-methylcoumarin hydrochloride, 62480-44-8, MFCD00043262, 50 mg
SDP
Catalog No. 50596416
Encompass_Preferred
MF: C16H20N2O3 . HCl, MW: 324.8, ≥99%, Appearance: White to white with light yellow cast powder. Fluorogenic substrate for leucine aminopeptidase. Sensitivity of fluorescence detection is greatly increased over other methods such as 4-Methoxy-naphthylamide (pNA) assays.
Catalog No. 50-596-416 Supplier AdipoGen Supplier No. CDXL0003M050
MF: C16H20N2O3 . HCl, MW: 324.8, ≥99%, Appearance: White to white with light yellow cast powder. Fluorogenic substrate for leucine aminopeptidase. Sensitivity of fluorescence detection is greatly increased over other methods such as 4-Methoxy-naphthylamide (pNA) assays.