Learn More
Medchemexpress LLC L-Leucine-d10 | 106972-44-5 | 141.23 | 100 MG
SDP
Catalog No. 5000390039
Encompass_Preferred
L-Leucine-d10 is a deuterium-labeled version of L-Leucine, an essential branched-chain amino acid (BCAA) known to activate the mTOR signaling pathway.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90039 Supplier Medchemexpress LLC Supplier No. HYN0486S100MG
L-Leucine-d10 is a deuterium-labeled version of L-Leucine, an essential branched-chain amino acid (BCAA) known to activate the mTOR signaling pathway.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.