Learn More
Medchemexpress LLC L-Leucine-d10 | 106972-44-5 | 141.23 | 25 MG
SDP
Catalog No. 5000389191
Encompass_Preferred
L-Leucine-d10 is deuterium-labeled L-Leucine. L-Leucine is an essential branched-chain amino acid (BCAA) that activates the mTOR signaling pathway. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89191 Supplier Medchemexpress LLC Supplier No. 25MGHYN0486S
Add to Cart
Add to Cart
L-Leucine-d10 is deuterium-labeled L-Leucine. L-Leucine is an essential branched-chain amino acid (BCAA) that activates the mTOR signaling pathway. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS