Learn More
Medchemexpress LLC L-leucine-d10 | 106972-44-5 | MFCD01321412 | 99.0% | 141.23 | C6H3D10NO2 | 10 MG
SDP
Catalog No. 5000296387
Encompass_Preferred
L-Leucine-d10 is the deuterium-labeled form of the essential branched-chain amino acid L-leucine, supplied as a high-purity research reagent for use as an isotopic tracer and internal standard in quantitative analyses (NMR, GC-MS, LC-MS) and metabolic tracing studies.
  • Deuterium-labeled (d10) L-leucine suitable for tracer studies.
  • High isotopic enrichment and 99.0% purity.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Supplied as a white to off-white solid in a 10 MG package.
  • Stable solid form for accurate quantitative measurements.
  • Store sealed at 4°C, protect from moisture and light.
Catalog No. 50-002-96387 Supplier Medchemexpress LLC Supplier No. HYN0486S10MG
L-Leucine-d10 is the deuterium-labeled form of the essential branched-chain amino acid L-leucine, supplied as a high-purity research reagent for use as an isotopic tracer and internal standard in quantitative analyses (NMR, GC-MS, LC-MS) and metabolic tracing studies.
  • Deuterium-labeled (d10) L-leucine suitable for tracer studies.
  • High isotopic enrichment and 99.0% purity.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Supplied as a white to off-white solid in a 10 MG package.
  • Stable solid form for accurate quantitative measurements.
  • Store sealed at 4°C, protect from moisture and light.