Learn More
Medchemexpress LLC L-Leucine-d2 | 362049-59-0 | 133.19 | 1 MG
SDP
Catalog No. 5000389792
Encompass_Preferred
L-Leucine-d2 is the deuterium labeled L-Leucine. L-Leucine is an essential branched-chain amino acid (BCAA), which activates the mTOR signaling pathway. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development. Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-89792 Supplier Medchemexpress LLC Supplier No. HYN0486S121MG
L-Leucine-d2 is the deuterium labeled L-Leucine. L-Leucine is an essential branched-chain amino acid (BCAA), which activates the mTOR signaling pathway. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development. Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs