Learn More
Medchemexpress LLC L-Leucine-d2 | 362049-59-0 | MFCD01861592 | 133.19 | 5 MG
SDP
Catalog No. 5000389874
Encompass_Preferred
L-Leucine-d2 is the deuterium-labeled form of L-Leucine, an essential branched-chain amino acid (BCAA) known to activate the mTOR signaling pathway. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer in quantitative analysis
  • Functions as an internal standard for various analytical methods
  • Activates the mTOR signaling pathway
  • Potential to affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-89874 Supplier Medchemexpress LLC Supplier No. HYN0486S125MG
L-Leucine-d2 is the deuterium-labeled form of L-Leucine, an essential branched-chain amino acid (BCAA) known to activate the mTOR signaling pathway. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer in quantitative analysis
  • Functions as an internal standard for various analytical methods
  • Activates the mTOR signaling pathway
  • Potential to affect pharmacokinetic and metabolic profiles