Learn More
Medchemexpress LLC L-Leucine-d3 | 87828-86-2 | 134.19 | 5 MG
SDP
Catalog No. 5000389375
Encompass_Preferred
L-Leucine-d3 is the deuterium labeled L-Leucine, an essential branched-chain amino acid (BCAA) that activates the mTOR signaling pathway. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during drug development. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89375 Supplier Medchemexpress LLC Supplier No. HYN0486S95MG
L-Leucine-d3 is the deuterium labeled L-Leucine, an essential branched-chain amino acid (BCAA) that activates the mTOR signaling pathway. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during drug development. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.